
Nateglinide Related Compound A

Synonym(s):
trans-4-Isopropylcyclohexanecarboxylic acid
Empirical Formula (Hill Notation):
C10H18O2
CAS Number:
Molecular Weight:
170.25
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2