
Naproxen Related Compound A

Synonym(s):
6-Methoxy-2-naphthoic acid, 6-Methoxy-2-naphthalenecarboxylic acid, 6-methoxynaphthalene-2-carboxylic acid
Linear Formula:
CH3OC10H6CO2H
CAS Number:
Molecular Weight:
202.21
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2