
Naproxen Impurity H

Synonym(s):
6-Methoxy-2-naphthol, 2-Hydroxy-6-methoxynaphthalene, 6-Methoxynaphthalen-2-ol
Empirical Formula (Hill Notation):
C11H10O2
CAS Number:
Molecular Weight:
174.20
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing