
Naloxone for peak identification

Synonym(s):
Naloxone hydrochloride dihydrate
Empirical Formula (Hill Notation):
C19H21NO4 · HCl · 2H2O
CAS Number:
Molecular Weight:
399.87
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing