Skip to Content
Merck
N-Allylthiourea 98%

N-Allylthiourea

N-Allylthiourea 98%
Synonym(s):
1-Allyl-2-thiourea, Thiosinamine, N-Allylthiourea
Linear Formula:
CH2=CHCH2NHCSNH2
CAS Number:
Molecular Weight:
116.18

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing