
N-(6-Chloro-3-pyridylmethyl)-N-cyano-N-methylacetamidine

Synonym(s):
Acetamiprid, sum of isomers
Empirical Formula (Hill Notation):
C10H11ClN4
CAS Number:
Molecular Weight:
222.67
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2