
Monostearyl maleate

Synonym(s):
Mono-Stearyl maleate
Empirical Formula (Hill Notation):
C22H40O4
CAS Number:
Molecular Weight:
368.55
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2