
Monoclonal Anti-Human IgG4 antibody produced in mouse

Synonym(s):
Monoclonal Anti-Human IgG4
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing