Skip to Content
Merck
Miconazole Related Compound I certified reference material, pharmaceutical secondary standard

Miconazole Related Compound I

Miconazole Related Compound I certified reference material, pharmaceutical secondary standard
Synonym(s):
1-{2-[(2-Chlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethyl}-1H-imidazole mononitrate
Empirical Formula (Hill Notation):
C18H15Cl3N2O · HNO3
CAS Number:
Molecular Weight:
444.70

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2