
Methyl linolenate

Synonym(s):
Linolenic acid methyl ester, Methyl cis,cis,cis-9,12,15-octadecatrienoate
Linear Formula:
CH3(CH2CH=CH)3(CH2)7COOCH3
CAS Number:
Molecular Weight:
292.46
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing