Skip to Content
Merck
Methyl butyrate analytical standard

Methyl butyrate

Methyl butyrate analytical standard
Linear Formula:
CH3CH2CH2COOCH3
CAS Number:
Molecular Weight:
102.13
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing