
Methyl behenate

Synonym(s):
Behenic acid methyl ester, Docosanoic acid methyl ester, Methyl docosanoate, Methyl behenate
Linear Formula:
CH3(CH2)20COOCH3
CAS Number:
Molecular Weight:
354.61
Products
Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3