Skip to Content
Merck
Mepiquat chloride PESTANAL®, analytical standard

Mepiquat chloride

Mepiquat chloride PESTANAL®, analytical standard
Synonym(s):
1,1-Dimethylpiperidinium chloride
Empirical Formula (Hill Notation):
C7H16ClN
CAS Number:
Molecular Weight:
149.66
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique