
L-Valine-13C5,15N

Synonym(s):
(S)-α-Aminoisovaleric acid-13C5,15N, L-2-Amino-3-methylbutanoic acid-13C5,15N, L-Valine-ul-13C,15N, 13C and 15N Labeled L-valine
Linear Formula:
(13CH3)213CH13CH(15NH2)13CO2H
CAS Number:
Molecular Weight:
123.10
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing