
Lithium ionophore IV

Synonym(s):
5-Butyl-5-ethyl-N,N,N′,N′-tetracyclohexyl-3,7-dioxaazelaic diamide, ETH 2137, N,N,N′,N′−Tetracyclohexyl(2-butyl-2-ethyltrimethylenedioxy)diacetamide
Empirical Formula (Hill Notation):
C37H66N2O4
CAS Number:
Molecular Weight:
602.93
Products
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing