
Liothyronine Sodium

Synonym(s):
(2S)-2-Amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoate sodium, 3,3′,5-Triiodo-L-thyronine sodium salt, Liothyronine, O-(4-Hydroxy-3-iodophenyl)-3,5-diiodo-L-tyrosine sodium salt, T3
Empirical Formula (Hill Notation):
C15H11I3NNaO4
CAS Number:
Molecular Weight:
672.96
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing