
L-Glutamic acid monosodium salt hydrate

Synonym(s):
(S)-2-Aminopentanedioic acid, L-Glutamic acid monosodium salt, Glu, MSG, Monosodium glutamate
Empirical Formula (Hill Notation):
C5H8NNaO4 · xH2O
CAS Number:
Molecular Weight:
169.11 (anhydrous basis)
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3