
Lactitol monohydrate

Synonym(s):
D-Lactitol monohydrate, 4-O-β-D-Galactopyranosyl-D-glucitol
Empirical Formula (Hill Notation):
C12H24O11 · H2O
CAS Number:
Molecular Weight:
362.33
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing