
Kaempferol 3-O-β -rutinoside

Synonym(s):
3,4′,5,7-Tetrahydroxyflavone 3-rhamnoglucoside, Nicotiflorin
Empirical Formula (Hill Notation):
C27H30O15
CAS Number:
Molecular Weight:
594.52
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing