Skip to Content
Merck
Iodomethane-13C,d3 99.5 atom % D, 99 atom % 13C, 99% (CP), contains copper as stabilizer

Iodomethane-13C,d3

Iodomethane-13C,d3 99.5 atom % D, 99 atom % 13C, 99% (CP), contains copper as stabilizer
Synonym(s):
Methyl-13C,d3 iodide
Linear Formula:
13CD3I
CAS Number:
Molecular Weight:
145.95

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Isotope
Application
Technique
Showing 1-1 of 1