
Hexane-d14

Synonym(s):
Tetradecadeuterohexane
Linear Formula:
CD3(CD2)4CD3
CAS Number:
Molecular Weight:
100.26
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Isotope
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing