Skip to Content
Merck
Hexachloro-1,3-butadiene analytical standard

Hexachloro-1,3-butadiene

Hexachloro-1,3-butadiene analytical standard
Synonym(s):
Perchlorobutadiene
Linear Formula:
CCl2=CClCCl=CCl2
CAS Number:
Molecular Weight:
260.76
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing