Hesperidin
Synonym(s):
3′,5,7-Trihydroxy 4′-methoxyflavanone 7-rutinoside, Hesperetin 7-rhamnoglucoside, Hesperitin-7-rutinoside, Hesperidin
Empirical Formula (Hill Notation):
C28H34O15
CAS Number:
Molecular Weight:
610.56
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing