
Guanosine 5′-triphosphate tris salt

Synonym(s):
GTP
Empirical Formula (Hill Notation):
C10H16N5O14P3 · 3 C4H11NO3
CAS Number:
Molecular Weight:
886.59
Products
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Tissue
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing