![Guanosine 5′-[β,γ-imido]triphosphate trisodium salt hydrate ≥85% (HPLC), powder](/deepweb/assets/sigmaaldrich/product/structures/204/494/05808804-1ca7-44bf-b6c5-d4934dc7cb85/640/05808804-1ca7-44bf-b6c5-d4934dc7cb85.png)
Guanosine 5′-[β,γ-imido]triphosphate trisodium salt hydrate
![Guanosine 5′-[β,γ-imido]triphosphate trisodium salt hydrate ≥85% (HPLC), powder](/deepweb/assets/sigmaaldrich/product/structures/204/494/05808804-1ca7-44bf-b6c5-d4934dc7cb85/640/05808804-1ca7-44bf-b6c5-d4934dc7cb85.png)
Synonym(s):
β:γ-Imidoguanosine 5′-triphosphate trisodium salt hydrate, 5′-Guanylyl-imidodiphosphate trisodium salt hydrate, GMP-PNP, Gpp(NH)p
Empirical Formula (Hill Notation):
C10H14N6Na3O13P3 · xH2O
CAS Number:
Molecular Weight:
588.14 (anhydrous basis)
Products
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing