
Guaiacol Impurity G

Synonym(s):
4-Methoxyphenol
Empirical Formula (Hill Notation):
C7H8O2
CAS Number:
Molecular Weight:
124.14
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing