Skip to Content
Merck
Geranic acid 85%, stabilised

Geranic acid

Geranic acid 85%, stabilised
Synonym(s):
3,7-Dimethyl-2,6-octadienoic acid
Linear Formula:
(CH3)2C=CHCH2CH2C(CH3)=CHCO2H
CAS Number:
Molecular Weight:
168.23
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing