Skip to Content
Merck
Fumaric acid European Pharmacopoeia (EP) Reference Standard

Fumaric acid

Fumaric acid European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Fumaric acid
Linear Formula:
HOOCCH=CHCOOH
CAS Number:
Molecular Weight:
116.07

Products

Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique