Skip to Content
Merck
Fosfomycin trometamol certified reference material, pharmaceutical secondary standard

Fosfomycin trometamol

Fosfomycin trometamol certified reference material, pharmaceutical secondary standard
Empirical Formula (Hill Notation):
C7H18NO7P
CAS Number:
Molecular Weight:
259.19

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing