
Fluoxetine impurity C

Synonym(s):
(±)-N-Methyl-(γ-[3-(trifluoromethyl)phenoxy]benzenepropanamine hydrochloride
Empirical Formula (Hill Notation):
C17H18F3NO · HCl
CAS Number:
Molecular Weight:
345.79
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing