
Fluazifop-P-butyl

Synonym(s):
Butyl (R)-2-[4-(5-trifluoromethyl-2-pyridyloxy)phenoxy]propionate
Empirical Formula (Hill Notation):
C19H20F3NO4
CAS Number:
Molecular Weight:
383.36
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing