
Flecainide acetate salt

Synonym(s):
N-(2-Piperidylmethyl)-2,5-bis-(2,2,2-trifluoroethoxy)benzamide acetate salt
Empirical Formula (Hill Notation):
C17H20F6N2O3 · C2H4O2
Products
Product Category
Brand
Formula Weight
Gene Alias
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing