Ethyl acetohydroxamate
Synonym(s):
Ethyl N-hydroxyacetimidate, Ethyl acetohydroximate
Linear Formula:
CH3C(=NOH)OC2H5
Products
Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing