
Ethinylestradiol for system suitability

Synonym(s):
17α-Ethynylestradiol, 17α-Ethynyl-1,3,5(10)-estratriene-3,17β-diol, 19-Nor-1,3,5(10),17α-pregnatrien-20-yne-3,17-diol, Ethinylestradiol
Empirical Formula (Hill Notation):
C20H24O2
CAS Number:
Molecular Weight:
296.40
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing