
Estradiol valerate for system suitability

Synonym(s):
β-Estradiol 17-valerate, 1,3,5(10)-Estratriene-3,17β-diol 17-pentanote
Empirical Formula (Hill Notation):
C23H32O3
CAS Number:
Molecular Weight:
356.50
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing