
Eosin Y

Synonym(s):
2′,4′,5′,7′-Tetrabromofluorescein, Acid Red 87, Bromo acid J. TS, XL, or XX, Bromofluorescein, Bronze Bromo ES, Eosin yellowish, Solvent red 43
Empirical Formula (Hill Notation):
C20H8Br4O5
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing