
Emtricitabine

Synonym(s):
(−)-2′3′-Dideoxy-5-fluoro-3′-thiacytidine, (2R,5S)-4-Amino-5-fluoro-1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2(1H)-one, FTC, Emtricitabine
Empirical Formula (Hill Notation):
C8H10FN3O3S
CAS Number:
Molecular Weight:
247.25
Products
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3