
Empty polypropylene SPE Tube with PE frits

Synonym(s):
Empty polypropylene SPE Tube with PE frits, 20 μm porosity
Products
Product Category
Brand
Volume (ml)
Application
Package Size
Pore size (µm)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing