![D-Val-Leu-Lys 4-nitroanilide dihydrochloride plasmin substrate](/deepweb/assets/sigmaaldrich/product/structures/212/901/9fa0533c-da08-4f4e-9569-6595cd80fede/640/9fa0533c-da08-4f4e-9569-6595cd80fede.png)
D-Val-Leu-Lys 4-nitroanilide dihydrochloride
![D-Val-Leu-Lys 4-nitroanilide dihydrochloride plasmin substrate](/deepweb/assets/sigmaaldrich/product/structures/212/901/9fa0533c-da08-4f4e-9569-6595cd80fede/640/9fa0533c-da08-4f4e-9569-6595cd80fede.png)
Synonym(s):
D-Val-Leu-Lys-pNA dihydrochloride, D-Valyl-L-leucyl-L-lysine 4-nitroanilide dihydrochloride
Empirical Formula (Hill Notation):
C23H38N6O5 · 2HCl
CAS Number:
Molecular Weight:
551.51
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing