
Doxycycline hydrochloride

Synonym(s):
2-Naphthacenecarboxamide, 4-(dimethylamino)- 1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methyl-1,11-dioxo-, hydrochloride (1:1), (4S,4aR,5S,5aR,6R,12aS)-
Empirical Formula (Hill Notation):
C22H24N2O8 · HCl
CAS Number:
Molecular Weight:
480.90
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing