
Dihexylammonium acetate solution

Synonym(s):
Dihexylamine acetate solution
Linear Formula:
[(CH3(CH2)5]2NH · CH3COOH
CAS Number:
Molecular Weight:
245.40
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing