
Dichloromethyl methyl ether

Synonym(s):
α,α−Dichloromethyl methyl ether, 1,1-Dichlorodimethyl ether, Dichloromethyl methyl ether
Linear Formula:
Cl2CHOCH3
CAS Number:
Molecular Weight:
114.96
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing