
Diammonium-15N2 hydrogen phosphate

Linear Formula:
(15NH4)2HPO4
CAS Number:
Molecular Weight:
134.04
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Isotope
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing