
D-Glucose-13C6,1,2,3,4,5,6,6-d7

Synonym(s):
D-Glucose-13C6,C-d7, Dextrose-13C6,C-d7
Empirical Formula (Hill Notation):
13C6H5D7O6
CAS Number:
Molecular Weight:
193.15
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing