Skip to Content
Merck
Desethylchloroquine ≥95% (HPLC), research grade

Desethylchloroquine

Desethylchloroquine ≥95% (HPLC), research grade
Synonym(s):
7-Chloro-4-{[4-(ethylamino)-1-methylbutyl]amino}quinoline, Monodesethylchloroqu​ine
Empirical Formula (Hill Notation):
C16H22ClN3
CAS Number:
Molecular Weight:
291.82

Products

Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing