(+)-Bicuculline
Synonym(s):
(+)-Bicuculline, Bucuculline
Empirical Formula (Hill Notation):
C20H17NO6
CAS Number:
Molecular Weight:
367.35
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing