Skip to Content
Merck
Cuminaldehyde ≥98%, FCC, FG

Cuminaldehyde

Cuminaldehyde ≥98%, FCC, FG
Synonym(s):
4-Isopropylbenzaldehyde
Linear Formula:
(CH3)2CHC6H4CHO
CAS Number:
Molecular Weight:
148.20
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Chemical Composition
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing