
C-Peptide solution

Empirical Formula (Hill Notation):
C129H211N35O48
CAS Number:
Molecular Weight:
3020.26
Products
Product Category
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing