Chlorhexidine
Synonym(s):
1,1′-Hexamethylenebis[5-(4-chlorophenyl)biguanide], Chlorhexidine
Linear Formula:
[-(CH2)3NHC(=NH)NHC(=NH)NHC6H4Cl]2
CAS Number:
Molecular Weight:
505.45
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Special Grade
Technique
Showing 1-4 of 4