
Cellulose Chromatography Resin

Empirical Formula (Hill Notation):
C116H116O64
CAS Number:
Molecular Weight:
2534.12
Products
Product Category
Formula Weight
Physical Form
Separation Technique
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing